Tisk…
Cut resistant gloves, PU800
Dodavatel: Marigold (Ansell)
1872E01000 1872E02000 1872E03000 1872E04000
MARG1872E01000EA 0 CZK
MARG1872E01000 MARG1872E02000 MARG1872E03000 MARG1872E04000
Cut resistant gloves, PU800
Rukavice
This seamless polyethylene High Tenacity/polyester knitted liner features a PU coating on the palm. It provides excellent abrasion and blade cuts resistance.
  • Fine and stretchable stitches for excellent comfort and adaptability
  • Latex knit wrist to ensure snug fit
  • High flexibility and dexterity for an optimum comfort in use
Ideal for fine tactile handling, sharp components handling, fitting and assembling works, quality control, automotive industry and glass industry.
EN 388 (4342), Cat. II
Certifikace: EN 388 (4342), Cat. II