Tisk…
ESD occupational shoes, clogs, Sandalen AGO, Roding, 11008
Dodavatel: SCHURR SCHUHVERTRIEB
1.1008.02.5239 1.1008.02.5240
SCRR1.1008.02.5239EA 0 CZK
SCRR1.1008.02.5239 SCRR1.1008.02.5240
ESD occupational shoes, clogs, Sandalen AGO, Roding, 11008
Obuv
Occupational shoe made from smooth leather, PU coated lining and a light slip-resistant PU rubber profile Manhatten sole.
  • Removable insole made from PUR-foam
For ESD and kitchen use.
Colour: Black
Width: 9
Weight: 355 g
Certifikace: EN 61340 (ESD); EN ISO 20347:2012; OB, SRA