Tisk…
T-shirts, A-Code® Basecamp
Dodavatel: FRISTADS KANSAS
1002413312XL 100241331L 100241331M 100241331S 100241331XL 1002415302XL 100241530L 100241530M 100241530S 100241530XL 100241540L 1002419002XL 100241900L 100241900M 100241900S 100241900XL 1002419102XL 100241910L 100241910M 100241910S 100241910XL 1002419402XL 100241940L 100241940M 100241940S 100241940XL
113-8415EA 209 CZK
113-8415 113-8413 113-8412 113-8411 113-8414 113-8410 113-8408 113-8407 113-8406 113-8409 113-8403 113-8400 113-8398 113-8397 113-8396 113-8399 113-8395 113-8393 113-8392 113-8391 113-8394 113-8390 113-8388 113-8387 113-8386 113-8389
T-shirts, A-Code® Basecamp
Pracovní oděvy
Basic T-shirts made of 100% pre-shrunk, ring spun cotton (single jersey). The models in mottled grey are made of 85% cotton and 15% viscose.
  • V-neck
  • Tubular construction, no side-seams
  • 2-ply rib with elastane
Weight: 150 g/m²
Certifikace: OEKO-TEX® certified
Objednat
Produktová specifikace
Model Basecamp, V-neck
Environmentally Preferable
Aplikace oblečení Casual
Risk area
Rod Pro muže
Sleeve or leg length Short
Tkanina 85% cotton and 15% viscose
Hmotnost 150 g/m²