Tisk…
Trousers, ladies, Fristads® FAS-253K
Dodavatel: FRISTADS KANSAS
10028154134 10028154136 10028154138 10028154140 10028154142 10028154144 10028154146 10028154148 10028194034 10028194036 10028194038 10028194040 10028194042 10028194044 10028194046 10028194048
KWIN10028154134EA 2940 CZK
KWIN10028154134 KWIN10028154136 KWIN10028154138 KWIN10028154140 KWIN10028154142 KWIN10028154144 KWIN10028154146 KWIN10028154148 KWIN10028194034 KWIN10028194036 KWIN10028194038 KWIN10028194040 KWIN10028194042 KWIN10028194044 KWIN10028194046 KWIN10028194048
Trousers, ladies, Fristads® FAS-253K
Pracovní oděvy
Work trousers made from FAS®, Fristads best marine twill made from 100% cotton. The knee and nail pockets are reinforced with Cordura®.
  • Two free-hanging, detachable front pockets – one with an extra compartment, the others with 3 smaller compartments and tool loops; nail pockets
  • Two front and 2 back pockets, extra roomy
  • Folding-rule pocket; pen pocket; phone pocket; tool pocket; leg pocket with flap
  • Two hammer loops, one of them adjustable
  • Button and loop for bladed tools
Material weight: 375 g/m²
Informace pro objednávání: Can be combined with:More Product Information
Objednat
Produktová specifikace
Model FAS-253K
Environmentally Preferable
Aplikace oblečení General Purpose,Industrial
Risk area
Rod Pro ženy
Sleeve or leg length Long
Typ uzávěru S knoflíky
Tkanina Cotton
Tloušťka 25 mm
Hmotnost 375 g/m²