Tisk…
Chlortetracycline hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
Nebezpečí
Synonyms: Chlortetracycline HCI, Aureomycin, 7-Chlorotetracycline hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride
M-Clarity™ quality level = MQ200
C4881-100G C4881-25G C4881-5G
SIALC4881-100GEA 14750 CZK
SIALC4881-100G SIALC4881-25G SIALC4881-5G
Chlortetracycline hydrochloride, Sigma-Aldrich®
Chlortetracycline hydrochloride
Vzorec: C₂₂H₂₃ClN₂O₈·HCl
MW: 515.34 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26142699
CAS číslo: 64-72-2
EINECS: 200-591-7