tert-Butyl isocyanide ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
t-Butylisocyanide
L19747.03
L19747.06
L19747.03EA
1450
CZK
L19747.03
L19747.06
tert-Butyl isocyanide ≥98%
tert-Butyl isocyanide
Purity: 98%
CAS No.: 7188-38-7
Molecular Formula: C5H9N
Formula Weight: 83.13
Storage Temperature: -30°C to -10°C
Flash Point: -2°(28°F)
Refractive Index: 1.3755
Odour: Stench
Boiling Point: 90-92°
Beilstein Reference: 1903156
UN No.: 1992
MDL No.: MFCD00000002
Vzorec:
C₅H₉N Bod varu: 90…92 °C Hustota: 0,735 Bod vzplanutí: –2 °C (28 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00000002 CAS číslo: 7188-38-7 EINECS: 230-553-5 UN: 1992 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...