Tisk…

3-Amino-1H-1,2,4-triazole-5-thiol ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-Amino-5-mercapto-1,2,4-triazole, 3-Amino-1,2,4-triazole-5-thiol, 3-Amino-4H-1,2,4-triazole-5-thiol, 3-Amino-5-mercapto-1,2,4-triazole 5-Amino-4H-1,2,4-triazole-3-thiol

A15242.06 A15242.14 A15242.22
A15242.06EA 982 CZK
A15242.06 A15242.14 A15242.22
3-Amino-1H-1,2,4-triazole-5-thiol ≥98%
3-Amino-1H-1,2,4-triazole-5-thiol

Purity: 98+% CAS No.: 16691-43-3 Molecular Formula: C2H4N4S Formula Weight: 116.15 Odour: Odourless Melting Point: 300° dec. Beilstein Reference: 112404 MDL No.: MFCD00005231

Vzorec: C₂H₄N₄S
MW: 116,15 g/mol
Bod tání: >300 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00005231
CAS číslo: 16691-43-3
EINECS: 240-735-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR