Kanamycin sulphate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate
M-Clarity™ quality level = MQ200
K1377-1G
K1377-25G
K1377-5G
SIALK1377-1GEA
2310
CZK
SIALK1377-1G
SIALK1377-25G
SIALK1377-5G
Kanamycin sulphate, Sigma-Aldrich®
Kanamycin sulphate
Vzorec:
C₁₈H₃₈N₄O₁₅S MW: 582.6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070253 CAS číslo: 25389-94-0 EINECS: 246-933-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...