5-Bromo-1,4-benzodioxane 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
5-Bromo-2,3-dihydro-1,4-benzodioxane
H27709.MD
H27709.03
H27709.MDEA
2450
CZK
H27709.MD
H27709.03
5-Bromo-1,4-benzodioxane 95%
5-Bromo-1,4-benzodioxane
Purity: 95%
CAS No.: 58328-39-5
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Refractive Index: 1.5955
Vzorec:
C₈H₇BrO₂ MW: 215,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03095029 CAS číslo: 58328-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...