Tisk…

Dichloroisocyanuric acid sodium salt monohydrate 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Troclosene sodium hydrate, Sodium dichloroisocyanurate hydrate

436450010 436452500 436450500
ACRO436450010EA 6000 CZK
ACRO436450010 ACRO436452500 ACRO436450500
Dichloroisocyanuric acid sodium salt monohydrate 97%
Dichloroisocyanuric acid sodium salt monohydrate
Vzorec: C₃Cl₂N₃NaO₃.H₂O
MW: 255,98 g/mol
Bod tání: 240…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00149404
CAS číslo: 52671-45-1
EINECS: 220-767-7
UN: 2465
ADR: 5.1,II

Objednat

Výsledky specifikačního testu

Appearance White Powder or crystals or granules
Infrared spectrum Conforms
Titration Iodometric 96.0 to 104.0 % (on dry substance)
Loss on drying 6 % to 11 % (105°C, 4 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR