Tisk…
Chemical resistant gloves, Showa 892
Dodavatel: Showa
892-08 892-09 892-10 892-07 892-11
112-0176EA 8590 CZK
112-0176 112-0177 112-0178 112-0175 112-0179
Chemical resistant gloves, Showa 892
Rukavice
These gloves with rolled cuffs and smooth finish are made of unsupported Viton and a butyl coating. They protect the hands from a wide array of chemical hazards.
  • Impermeable for working in damp or greasy environments, e.g. petrochemical industries
  • Thin, light glove that feels like a 'second skin'
  • Cuff prevents dirt from entering the glove and protects the wrist
Thickness: 0,3 mm
Colour: Black
EN 388 (2000), EN 374 (DFL); Cat. III
Certifikace: EN 388:2016 (1010X), EN ISO 374-1 Type B (DFL), EN ISO 374-5 VIRUS; Cat. III