Tisk…

3-Thienylacetic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Thiophene-3-acetic acid, 3-Thiopheneacetic acid

M-Clarity™ quality level = MQ100

89017-25G 89017-5G
SIAL89017-25GEA 8660 CZK
SIAL89017-25G SIAL89017-5G
3-Thienylacetic acid, Sigma-Aldrich®
3-Thienylacetic acid
Vzorec: C₆H₆O₂S
MW: 142,18 g/mol
MDL Number: MFCD00005473
CAS číslo: 6964-21-2
EINECS: 230-166-1
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR