Tisk…

3-Thienylacetic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A10700.06 A10700.14 A10700.22
A10700.06EA 1880 CZK
A10700.06 A10700.14 A10700.22
3-Thienylacetic acid ≥98%
3-Thienylacetic acid

Purity: 98% CAS No.: 6964-21-2 Molecular Formula: C6H6O2S Formula Weight: 142.18 Melting Point: 77-81° Boiling Point: 129°/2.5mm UN No.: 3261 MDL No.: MFCD00005473

Vzorec: C₆H₆O₂S
Bod varu: 129 °C (2,5 mmHg)
Bod tání: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00005473
CAS číslo: 6964-21-2
EINECS: 230-166-1
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR