2-Thienylacetic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-Thiopheneacetic acid, Thiophene-2-acetic acid, 2-(Thiophen-2-yl)acetic acid
A10394.14
A10394.22
A10394.36
A10394.14EA
1720
CZK
A10394.14
A10394.22
A10394.36
2-Thienylacetic acid ≥98%
2-Thienylacetic acid
Purity: 98%
CAS No.: 1918-77-0
Molecular Formula: C6H6O2S
Formula Weight: 142.18
Melting Point: 61-65°
Boiling Point: 160°/22mm
UN No.: 3261
MDL No.: MFCD00005458
Vzorec:
C₆H₆O₂S Bod varu: 160 °C (22 mmHg) Bod tání: 61…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005458 CAS číslo: 1918-77-0 EINECS: 217-639-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...