Tisk…

Cyanamide ≥98% stabilizovaný

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Stabilization: Stabilised with phosphoric acid

L03744.06 L03744.14 L03744.22
L03744.06EA 1040 CZK
L03744.06 L03744.14 L03744.22
Cyanamide ≥98% stabilizovaný
Cyanamide

Stabilized


Purity: 98+% CAS No.: 420-04-2 Molecular Formula: CH2N2 Formula Weight: 42.04 Storage Temperature: 2°C to 8°C Flash Point: 141°(285°F) Odour: Odourless pH: 3.5-4.2 Melting Point: 43-47° Boiling Point: 132-138°/12mm Beilstein Reference: 1732569 Merck Reference: 14,2684 UN No.: 2923 MDL No.: MFCD00007572

Vzorec: CH₂N₂
Bod varu: 132…138 °C (12 mmHg)
Bod tání: 43…47 °C
Hustota: 1,282
Bod vzplanutí: 141 °C (285 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007572
CAS číslo: 420-04-2
EINECS: 206-992-3
UN: 2923
ADR: 8,III
Merck Index: 14,2684

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR