Orotic acid monohydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Orotic acid hydrate, 2,6-Dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate
M-Clarity™ quality level = MQ200
O8402-100G
O8402-25G
SIALO8402-100GEA
2680
CZK
SIALO8402-100G
SIALO8402-25G
Orotic acid monohydrate, Sigma-Aldrich®
Orotic acid monohydrate
Vzorec:
C₅H₄N₂O₄·1H₂O |
MDL Number:
MFCD00149398 CAS číslo: 50887-69-9 EINECS: 200-619-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...