Tisk…

Abietic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

00010-100G 00010-25G
SIAL00010-100GEA 0 CZK
SIAL00010-100G SIAL00010-25G
Abietic acid, Sigma-Aldrich®
Abietic acid
Vzorec: C₂₀H₃₀O₂
MW: 302,45 g/mol
MDL Number: MFCD03423567
CAS číslo: 514-10-3
EINECS: 208-178-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR