Tisk…

(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: L-Lactide, (3S,6S)-3,6-Dimethyl-1,4-dioxane-2,5-dione

M-Clarity™ quality level = MQ100

367044-25G 367044-100G
SIAL367044-25GEA 2560 CZK
SIAL367044-25G SIAL367044-100G
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione, Sigma-Aldrich®
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione
Vzorec: C₆H₈O₄
MW: 144,13 g/mol
MDL Number: MFCD00070594
CAS číslo: 4511-42-6
EINECS: 224-832-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR