Tisk…

(S)-(-)-Indoline-2-carboxylic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

346802-1G 346802-5G
SIAL346802-1GEA 1210 CZK
SIAL346802-1G SIAL346802-5G
(S)-(-)-Indoline-2-carboxylic acid, Sigma-Aldrich®
(S)-(-)-Indoline-2-carboxylic acid
Vzorec: C₉H₉NO₂
MDL Number: MFCD00070578
CAS číslo: 79815-20-6
EINECS: 410-860-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR