Phenylmagnesium chloride 2 mol/l in THF, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
224448-100ML
224448-800ML
224448-8L
SIAL224448-100MLEA
2860
CZK
SIAL224448-100ML
SIAL224448-800ML
SIAL224448-8L
Phenylmagnesium chloride 2 mol/l in THF, Sigma-Aldrich®
Phenylmagnesium chloride
Vzorec:
C₆H₅ClMg MW: 136,86 g/mol |
MDL Number:
MFCD00000464 CAS číslo: 100-59-4 EINECS: 202-868-8 UN: 3399 ADR: 4.3,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...