Tisk…
Phenylmagnesium bromide 3 mol/l in diethyl ether, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
171565-100ML 171565-50ML 171565-800ML
SIAL171565-100MLEA 1870 CZK
SIAL171565-100ML SIAL171565-50ML SIAL171565-800ML
Phenylmagnesium bromide 3 mol/l in diethyl ether, Sigma-Aldrich®
Phenylmagnesium bromide
Vzorec: C₆H₅BrMg
MDL Number: MFCD00000038
CAS číslo: 100-58-3
EINECS: 202-867-2
UN: 3399
ADR: 4.3,II