Tisk…

DL-Menthol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol

M-Clarity™ quality level = MQ300

W266507-1KG-K W266507-250G-K W266507-4KG-K
SIALW266507-1KG-KEA 3980 CZK
SIALW266507-1KG-K SIALW266507-250G-K SIALW266507-4KG-K
DL-Menthol, Sigma-Aldrich®
DL-Menthol
Vzorec: C₁₀H₂₀O
MW: 156,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001484
CAS číslo: 89-78-1
EINECS: 201-939-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR