Tisk…

DL-Menthol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: rel-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol

63670-100G-F 63670-500G-F
SIAL63670-100G-FEA 1430 CZK
SIAL63670-100G-F SIAL63670-500G-F
DL-Menthol, Sigma-Aldrich®
DL-Menthol
Vzorec: C₁₀H₂₀O
MW: 156,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001484
CAS číslo: 89-78-1
EINECS: 201-939-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR