Tisk…

β-Cyclodextrine hydrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

856088-25G 856088-100G 856088-5G
SIAL856088-25GEA 1690 CZK
SIAL856088-25G SIAL856088-100G SIAL856088-5G
β-Cyclodextrine hydrate, Sigma-Aldrich®
β-Cyclodextrine hydrate
Vzorec: C₄₂H₇₀O₃₅·H₂O
MDL Number: MFCD00150811
CAS číslo: 68168-23-0
EINECS: 231-493-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR