2,5-Dibromo-3,4-dinitrothiophene 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L10348.03
L10348.06
L10348.03EA
2080
CZK
L10348.03
L10348.06
2,5-Dibromo-3,4-dinitrothiophene 95%
2,5-Dibromo-3,4-dinitrothiophene
Purity: 95%
CAS No.: 52431-30-8
Molecular Formula: C4Br2N2O4S
Formula Weight: 331.94
Melting Point: 134-137°
Beilstein Reference: 218219
Vzorec:
C₄Br₂N₂O₄S MW: 331.94 g/mol Bod tání: 134…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015537 CAS číslo: 52431-30-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...