2-Iodo-5-nitropyridine ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H27103.03
H27103.06
H27103.03EA
2140
CZK
H27103.03
H27103.06
2-Iodo-5-nitropyridine ≥97%
2-Iodo-5-nitropyridine
Purity: 97%
CAS No.: 28080-54-8
Molecular Formula: C5H3IN2O2
Formula Weight: 249.98
Melting Point: 163-168°
MDL No.: MFCD05863241
Vzorec:
C₅H₃IN₂O₂ Bod tání: 163…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD05863241 CAS číslo: 28080-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...