1,8-Bis(dimethylamino)naphthalene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
14795-10G
14795-50G
SIAL14795-10GEA
1560
CZK
SIAL14795-10G
SIAL14795-50G
1,8-Bis(dimethylamino)naphthalene, Sigma-Aldrich®
1,8-Bis(dimethylamino)naphthalene
Vzorec:
C₁₄H₁₈N₂ |
MDL Number:
MFCD00003920 CAS číslo: 20734-58-1 EINECS: 244-001-6 UN: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...