Tisk…

1-Chloroisoquinoline, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

156744-1G 156744-5G
SIAL156744-1GEA 1630 CZK
SIAL156744-1G SIAL156744-5G
1-Chloroisoquinoline, Sigma-Aldrich®
1-Chloroisoquinoline
Vzorec: C₉H₆ClN
MW: 163,61 g/mol
MDL Number: MFCD00024134
CAS číslo: 19493-44-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR