(S)-(+)-Prolinol ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(S)-(+)-2-Hydroxymethylpyrrolidine, (2S)-pyrrolidin-2-ylmethanol, L(+)-Prolinol, S(+)-2-Pyrrolidinemethanol, L-Prolinol
L09779.03
L09779.06
L09779.14
L09779.03EA
1350
CZK
L09779.03
L09779.06
L09779.14
(S)-(+)-Prolinol ≥98%
(S)-(+)-Prolinol
Purity: 98%
CAS No.: 23356-96-9
Molecular Formula: C5H11NO
Formula Weight: 101.15
Flash Point: 86°(186°F)
Refractive Index: 1.4853
Melting Point: 42-44°
Boiling Point: 74-76°/2mm
Beilstein Reference: 79843
Optical Rotation: +31° (c=1 in toluene)
MDL No.: MFCD00005255
Vzorec:
C₅H₁₁NO MW: 101,15 g/mol Bod varu: 74…76 °C (2 mmHg) Bod tání: 42…44 °C Hustota: 1,025 Bod vzplanutí: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005255 CAS číslo: 23356-96-9 EINECS: 245-605-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...