(R)-(-)-Prolinol ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
D-Prolinol, (2R)-pyrrolidin-2-ylmethanol, D(-)-Prolinol
L11109.03
L11109.06
L11109.14
L11109.03EA
1330
CZK
L11109.03
L11109.06
L11109.14
(R)-(-)-Prolinol ≥98%
(R)-(-)-Prolinol
Purity: 98+%
CAS No.: 68832-13-3
Molecular Formula: C5H11NO
Formula Weight: 101.15
Flash Point: 86°(186°F)
Refractive Index: 1.4849
Boiling Point: 74-76°/2mm
Beilstein Reference: 1523669
Optical Rotation: -31° (c=1 in toluene)
MDL No.: MFCD00064321
Vzorec:
C₅H₁₁NO MW: 101,1 g/mol Bod varu: 74…76 °C (2 mmHg) Hustota: 1,025 Bod vzplanutí: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064321 CAS číslo: 68832-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...