Tisk…

4-Acetamido-2-methylbenzoic acid 96%

Dodavatel: Thermo Fisher Scientific
Varování

H27174.14 H27174.06
H27174.14EA 5630 CZK
H27174.14 H27174.06
4-Acetamido-2-methylbenzoic acid 96%
4-Acetamido-2-methylbenzoic acid

Purity: 96% CAS No.: 103204-69-9 Molecular Formula: C10H11NO3 Formula Weight: 193.21 Melting Point: ca 248° dec. MDL No.: MFCD02258874

Vzorec: C₁₀H₁₁NO₃
MW: 193.21 g/mol
Bod tání: ∼ 248 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD02258874
CAS číslo: 103204-69-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR