3-(Methoxycarbonyl)phenylboronic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Methyl 3-boronobenzoate, 3-(Methoxycarbonyl)benzeneboronic acid
M-Clarity™ quality level = MQ100
591130-1G
591130-5G
SIAL591130-1GEA
2390
CZK
SIAL591130-1G
SIAL591130-5G
3-(Methoxycarbonyl)phenylboronic acid, Sigma-Aldrich®
3-(Methoxycarbonyl)phenylboronic acid
Vzorec:
C₈H₉BO₄ |
MDL Number:
MFCD02093046 CAS číslo: 99769-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...