2-(Methoxycarbonyl)phenylboronic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Methoxycarbonylphenylboronic acid, 2-(Methoxycarbonyl)benzeneboronic acid
L17958.03
L17958.06
L17958.03EA
1170
CZK
L17958.03
L17958.06
2-(Methoxycarbonyl)phenylboronic acid ≥97%
2-(Methoxycarbonyl)phenylboronic acid
Purity: 97%
CAS No.: 374538-03-1
Molecular Formula: C8H9BO4
Formula Weight: 179.97
Melting Point: 68-70°
Beilstein Reference: 8313459
Vzorec:
C₈H₉BO₄ MW: 179,97 g/mol Bod tání: 68…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179452 CAS číslo: 374538-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...