5-Iodo-1,2,3-trimethoxybenzene ≥98%
Dodavatel: Thermo Scientific
L17414.03
L17414.06
L17414.03EA
2320
CZK
L17414.03
L17414.06
5-Iodo-1,2,3-trimethoxybenzene ≥98%
5-Iodo-1,2,3-trimethoxybenzene
Purity: 98+%
CAS No.: 25245-29-8
Molecular Formula: C9H11IO3
Formula Weight: 294.09
Melting Point: 85-87°
Beilstein Reference: 1529690
Vzorec:
C₉H₁₁IO₃ MW: 294,09 g/mol Bod tání: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318153 CAS číslo: 25245-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...