Tisk…

5-Acetamido-2-aminobenzoic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

L11256.06 L11256.14
L11256.06EA 1290 CZK
L11256.06 L11256.14
5-Acetamido-2-aminobenzoic acid 97%
5-Acetamido-2-aminobenzoic acid

Purity: 97% CAS No.: 50670-83-2 Molecular Formula: C9H10N2O3 Formula Weight: 194.19 Melting Point: ca 230° dec. Beilstein Reference: 2726882

Vzorec: C₉H₁₀N₂O₃
MW: 194.19 g/mol
Bod tání: ∼ 230 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00060120
CAS číslo: 50670-83-2
EINECS: 256-702-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR