Tisk…

4-(Allyloxy)benzaldehyde ≥97%

Dodavatel: Thermo Scientific
Varování

A13773.09 A13773.18
A13773.09EA 1760 CZK
A13773.09 A13773.18
4-(Allyloxy)benzaldehyde ≥97%
4-(Allyloxy)benzaldehyde

Purity: 97% CAS No.: 40663-68-1 Molecular Formula: C10H10O2 Formula Weight: 162.19 Flash Point: 110°(230°F) Refractive Index: 1.5695 Boiling Point: 150-152°/18mm Beilstein Reference: 2206924 MDL No.: MFCD00014133

Vzorec: C₁₀H₁₀O₂
MW: 162,19 g/mol
Bod varu: 150…152 °C (18 mmHg)
Hustota: 1,058
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014133
CAS číslo: 40663-68-1
EINECS: 255-027-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR