4-Bromophenyl phenyl ether (BDE No. 3), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
B65209-100G
B65209-25G
SIALB65209-100GEA
4300
CZK
SIALB65209-100G
SIALB65209-25G
4-Bromophenyl phenyl ether (BDE No. 3), Sigma-Aldrich®
4-Bromophenyl phenyl ether (BDE No. 3)
Vzorec:
C₁₂H₉BrO |
MDL Number:
MFCD00000094 CAS číslo: 101-55-3 EINECS: 202-952-4 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...