Tisk…

3-(Trimethoxysilyl)propyl methacrylate ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: γ-methacryloxypropyl-trimethoxysilane, 3-(Methacryloyloxy)propyltrimethoxysilane

A17714.22 A17714.36 AB117674.0100
A17714.22EA 1560 CZK
A17714.22 A17714.36 ABCRAB117674.0100
3-(Trimethoxysilyl)propyl methacrylate ≥97%
3-(Trimethoxysilyl)propyl methacrylate

Purity: 97% CAS No.: 2530-85-0 Molecular Formula: C10H20O5Si Formula Weight: 248.35 Flash Point: 92°(197°F) Refractive Index: 1.431 Boiling Point: 190° Beilstein Reference: 1952435 MDL No.: MFCD00008593

Vzorec: C₁₀H₂₀O₅Si
Bod varu: 190 °C
Hustota: 1,045
Bod vzplanutí: 92 °C (197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008593
CAS číslo: 2530-85-0
EINECS: 219-785-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR