Tisk…

Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: H-Gly-OBzl·HCl, H-DL-Gly-OBzl hydrochloride, Benzyl glycinate hydrochloride, H-DL-Gly-OBzl.HCl

M-Clarity™ quality level = MQ200

G3267-1G G3267-25G G3267-5G
SIALG3267-1GEA 1930 CZK
SIALG3267-1G SIALG3267-25G SIALG3267-5G
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl), Sigma-Aldrich®
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl)
Vzorec: C₉H₁₂ClNO₂
MW: 201,65 g/mol
MDL Number: MFCD00035442
CAS číslo: 2462-31-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR