Tisk…

3,6-Dioxaoctamethylenediamine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

385506-100ML 385506-500ML
SIAL385506-100MLEA 2750 CZK
SIAL385506-100ML SIAL385506-500ML
3,6-Dioxaoctamethylenediamine, Sigma-Aldrich®
3,6-Dioxaoctamethylenediamine
Vzorec: C₆H₁₆N₂O₂
MDL Number: MFCD00040474
CAS číslo: 929-59-9
EINECS: 213-203-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR