3,6-Dioxaoctamethylenediamine ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
44759.09
44759.18
44759.30
44759.09EA
1370
CZK
44759.09
44759.18
44759.30
3,6-Dioxaoctamethylenediamine ≥97%
3,6-Dioxaoctamethylenediamine
Purity: 97+%
CAS No.: 929-59-9
Molecular Formula: (CH2OCH2CH2NH2)2
Formula Weight: 148.20
Flash Point: 121°(250°F)
Physical Form: Liquid
Density: 1.01
Refractive Index: 1.4610
Sensitivity: Hygroscopic
Boiling Point: 105-109°/6mm
Beilstein Reference: 1739187
UN No.: 2735
MDL No.: MFCD00040474
Vzorec:
C₆H₁₆N₂O₂ Bod varu: 105…109 °C (6 mmHg) Hustota: 1,01 Bod vzplanutí: 121 °C (250 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040474 CAS číslo: 929-59-9 EINECS: 213-203-6 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...