N,N,N',N',N''-Pentamethyldiethylenetriamine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine
M-Clarity™ quality level = MQ200
369497-250ML
369497-1L
SIAL369497-250MLEA
3700
CZK
SIAL369497-250ML
SIAL369497-1L
N,N,N',N',N''-Pentamethyldiethylenetriamine, Sigma-Aldrich®
N,N,N',N',N''-Pentamethyldiethylenetriamine
Vzorec:
C₉H₂₃N₃ MW: 173.30 g/mol |
MDL Number:
MFCD00014876 CAS číslo: 3030-47-5 EINECS: 221-201-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...