Tisk…

N,N,N',N',N''-Pentamethyldiethylenetriamine 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine

A15433.14 A15433.22 A15433.36
A15433.14EA 1660 CZK
A15433.14 A15433.22 A15433.36
N,N,N',N',N''-Pentamethyldiethylenetriamine 98%
N,N,N',N',N''-Pentamethyldiethylenetriamine

Purity: 98% CAS No.: 3030-47-5 Molecular Formula: C9H23N3 Formula Weight: 173.30 Flash Point: 72°(161°F) Refractive Index: 1.4420 Melting Point: -20° Boiling Point: 199° Beilstein Reference: 1741396 UN No.: 2922 MDL No.: MFCD00014876

Vzorec: C₉H₂₃N₃
MW: 173.30 g/mol
Bod varu: 199 °C
Bod tání: < –20 °C
Hustota: 0.830
Bod vzplanutí: 72 °C (161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014876
CAS číslo: 3030-47-5
EINECS: 221-201-1
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR