Tisk…

Triethyl methanetricarboxylate ≥98%

Dodavatel: Thermo Scientific

L08356.06 L08356.14
L08356.06EA 924 CZK
L08356.06 L08356.14
Triethyl methanetricarboxylate ≥98%
Triethyl methanetricarboxylate

Purity: 98% CAS No.: 6279-86-3 Molecular Formula: C10H16O6 Formula Weight: 232.23 Flash Point: 110°(230°F) Refractive Index: 1.4255 Melting Point: 26-29° Boiling Point: 253° Beilstein Reference: 1793645 MDL No.: MFCD00009150

Vzorec: C₁₀H₁₆O₆
MW: 232,23 g/mol
Bod varu: 253 °C
Bod tání: 26…29 °C
Hustota: 1,095
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009150
CAS číslo: 6279-86-3
EINECS: 228-477-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR