Tisk…

Triethyl methanetricarboxylate 98%

Dodavatel: Thermo Scientific

139630250 139631000
ACRO139630250EA 3870 CZK
ACRO139630250 ACRO139631000
Triethyl methanetricarboxylate 98%
Triethyl methanetricarboxylate
Vzorec: C₁₀H₁₆O₆
MW: 232,23 g/mol
Bod varu: 253 °C (760 mmHg)
Bod tání: 29 °C
Hustota: 1,1 g/cm³
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009150
CAS číslo: 6279-86-3
EINECS: 228-477-2

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4260 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR