Tisk…

Glycine propyl ester hydrochloride

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Glycine n-propyl ester hydrochloride

H50250.03 H50250.06
H50250.03EA 1220 CZK
H50250.03 H50250.06
Glycine propyl ester hydrochloride
Glycine propyl ester hydrochloride

CAS No.: 13049-01-9 Molecular Formula: H2NCH2CO2(CH2)2CH3.HCl Formula Weight: 117.15 Melting Point: 78-80° MDL No.: MFCD00151829

Vzorec: C₅H₁₂ClNO₂
MW: 117.15 g/mol
Bod tání: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00151829
CAS číslo: 13049-01-9
EINECS: 235-925-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR