Tisk…

Glycine iso-propyl ester hydrochloride (H-Gly-OiPr.HCl) 96%

Dodavatel: Thermo Scientific

Synonyms: H-Gly-OIpr·HCl, Isopropyl 2-aminoacetate hydrochloride

H50249.03 H50249.06
H50249.03EA 1180 CZK
H50249.03 H50249.06
Glycine iso-propyl ester hydrochloride (H-Gly-OiPr.HCl) 96%
Glycine iso-propyl ester hydrochloride (H-Gly-OiPr.HCl)

Purity: 96% CAS No.: 14019-62-6 Molecular Formula: C5H11NO2.HCl Formula Weight: 153.61 MDL No.: MFCD00151826

Vzorec: C₅H₁₂ClNO₂
MW: 153,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151826
CAS číslo: 14019-62-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR