Tisk…

Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: H-Gly-OtBu.HCl, Glycine tert-butyl ester hydrochloride, H-Gly-OtBu·HCl

M-Clarity™ quality level = MQ200

347957-1G 347957-5G
SIAL347957-1GEA 1290 CZK
SIAL347957-1G SIAL347957-5G
Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl), Sigma-Aldrich®
Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl)
Vzorec: C₆H₁₄ClNO₂
MW: 167,64 g/mol
MDL Number: MFCD00058255
CAS číslo: 27532-96-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR