Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl) ≥99%
Dodavatel: Thermo Fisher Scientific
L17116.03
L17116.06
L17116.03EA
927
CZK
L17116.03
L17116.06
Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl) ≥99%
Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl)
Purity: 99%
CAS No.: 27532-96-3
Molecular Formula: H2NCH2CO2C(CH3)3.HCl
Formula Weight: 167.64
Melting Point: ca 140° dec.
Beilstein Reference: 3989048
Vzorec:
C₆H₁₄ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00058255 CAS číslo: 27532-96-3 EINECS: 214-223-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...