Tisk…

1,3-Bis(trimethylsilyl)urea ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: N,N'-Bis(trimethylsilyl)urea

L01301.18 L01301.30
L01301.18EA 913 CZK
L01301.18 L01301.30
1,3-Bis(trimethylsilyl)urea ≥98%
1,3-Bis(trimethylsilyl)urea

Purity: 98+% CAS No.: 18297-63-7 Molecular Formula: C7H20N2OSi2 Formula Weight: 204.42 Melting Point: ca 230° dec. Beilstein Reference: 1937452

Vzorec: C₇H₂₀N₂OSi₂
Storage Temperature: Ambient
MDL Number: MFCD00008260
CAS číslo: 18297-63-7
EINECS: 242-177-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR